siRNA Details
Gene Symbol: Usp36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033528.1 | NP_001028700.1 | 6 | 1020 | ||
GenBank mRNA | AK028572.1 | - | 6 | 1065 | ||
AK049205.1 | - | 6 | 850 | |||
AK129363.1 | - | 6 | 818 |
