siRNA Details
Gene Symbol: Cdpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164625.1 | NP_001158097.1 | 5 | 551 | ||
NM_001164626.1 | NP_001158098.1 | 4 | 345 | |||
NM_197998.2 | NP_932115.2 | 5 | 492 | |||
GenBank mRNA | AK008763.1 | - | 2 | 179 | ||
BC025858.1 | - | 5 | 454 | 5007877 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
