siRNA Details
Gene Symbol: 2210408I21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081353.1 | NP_001074822.1 | 4 | 875 | ||
NM_001145676.1 | NP_001139148.1 | 4 | 875 | |||
GenBank mRNA | AK008859.1 | - | 4 | 863 | ||
AK143619.1 | - | 4 | 813 | |||
AK165312.1 | - | 4 | 872 | |||
BC089469.1 | - | 4 | 857 | |||
BC151066.1 | - | 4 | 857 | 9087979 | ||
BC157949.1 | - | 4 | 857 | 9088124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
