siRNA Details
Gene Symbol: Rbm12b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028226.2 | NP_082502.2 | 3 | 2810 | ||
GenBank mRNA | AK013872.1 | - | 1 | 1905 | ||
BC031713.1 | - | 1 | 2407 | |||
BC050844.1 | - | 3 | 2611 | 6513134 | ||
BC054081.1 | - | 1 | 1543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
