siRNA Details
Gene Symbol: Ssbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023672.2 | NP_076161.2 | 11 | 1142 | ||
NM_198438.1 | NP_940840.1 | 10 | 1061 | |||
GenBank mRNA | AF170906.1 | - | 10 | 791 | ||
AF500117.1 | - | 10 | 791 | |||
AK011509.1 | - | 11 | 760 | |||
AK011853.1 | - | 11 | 1112 | |||
AK017711.1 | - | 11 | 1146 | |||
AK041388.1 | - | 7 | 658 | |||
AK133812.1 | - | 8 | 595 | |||
AK153733.1 | - | 7 | 502 | |||
AY167987.1 | - | 10 | 674 | |||
BC003430.1 | - | 10 | 815 | 2646149 | ||
BC028324.1 | - | 5 | 283 | |||
BC035475.1 | - | 10 | 1004 |

More Information
Other Species Targeted

Gene Ontology Categories:
