siRNA Details
Gene Symbol: Taf4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100449.1 | NP_001093919.1 | 12 | 2491 | ||
GenBank mRNA | AK029943.1 | - | 1 | 36 |

More Information
Other Species Targeted

Gene Ontology Categories:
