siRNA Details
Gene Symbol: Exosc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028274.4 | NP_082550.1 | 1 | 1098 | ||
GenBank mRNA | AK012102.1 | - | 1 | 1097 | ||
AK088532.1 | - | 1 | 1099 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
