siRNA Details
Gene Symbol: Reep4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180588.2 | NP_850919.1 | 7 | 1052 | ||
GenBank mRNA | AK012301.1 | - | 7 | 949 | ||
AY562232.1 | - | 7 | 596 | |||
BC033929.1 | - | 7 | 680 | 1363539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
