siRNA Details
Gene Symbol: Zfp566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152814.2 | NP_690027.1 | 4 | 669 | ||
GenBank mRNA | AK012367.1 | - | 4 | 647 | ||
BC019790.1 | - | 4 | 556 | 5151196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
