siRNA Details
Gene Symbol: Uaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028283.2 | NP_082559.1 | 12 | 1098 | ||
GenBank mRNA | AB030647.1 | - | 12 | 1168 | ||
AK148900.1 | - | 12 | 1204 | |||
AK173203.1 | - | 1 | 994 | |||
BC033470.1 | - | 12 | 1015 | 4951775 | ||
BC042415.1 | - | 12 | 1098 | 4165488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
