siRNA Details
Gene Symbol: Lypd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145100.3 | NP_659568.2 | 2 | 421 | ||
GenBank mRNA | AB041649.1 | - | 2 | 421 | ||
AK012406.1 | - | 2 | 109 | |||
AK045592.1 | - | 2 | 422 | |||
AK082941.1 | - | 2 | 101 | |||
BC058599.1 | - | 2 | 354 | 6313510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
