siRNA Details
Gene Symbol: Lins | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191001.1 | NP_001177930.1 | 8 | 2302 | ||
NM_152815.2 | NP_690028.2 | 8 | 2317 | |||
GenBank mRNA | AB083158.1 | - | 6 | 1901 | ||
AK012554.1 | - | 8 | 2164 | |||
AK156066.1 | - | 8 | 2292 | |||
AK171170.1 | - | 8 | 2229 | |||
BC030943.1 | - | 10 | 2309 | |||
BC058648.1 | - | 8 | 2134 | |||
BC059228.1 | - | 3 | 1303 | |||
BC138549.1 | - | 6 | 1975 | 8861569 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
