siRNA Details
Gene Symbol: 2810006K23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134717.2 | NP_001128189.1 | 2 | 341 | ||
NM_028310.3 | NP_082586.1 | 2 | 341 | |||
GenBank mRNA | AK012682.1 | - | 2 | 241 | ||
AK039255.1 | - | 2 | 342 | |||
AK140664.1 | - | 2 | 284 | |||
BC046909.1 | - | 2 | 293 | 6466378 | ||
BC054073.1 | - | 2 | 293 |

More Information
Other Species Targeted

Gene Ontology Categories:
