siRNA Details
Gene Symbol: Dnajc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164583.1 | NP_001158055.1 | 19 | 2901 | ||
NM_001164584.1 | NP_001158056.1 | 19 | 2846 | |||
NM_001164585.1 | NP_001158057.1 | 20 | 2911 | |||
NM_198412.2 | NP_940804.1 | 20 | 2820 | |||
GenBank mRNA | AK049935.1 | - | 7 | 863 | ||
AK122293.1 | - | 19 | 2822 | |||
AK147570.1 | - | 20 | 2783 | |||
AK147637.1 | - | 20 | 2783 | |||
AK163657.1 | - | 19 | 2720 | |||
BC035262.1 | - | 14 | 2055 | |||
BC060734.1 | - | 19 | 2901 | 6409942 | ||
BC064460.1 | - | 20 | 2789 | 6821822 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
