siRNA Details
Gene Symbol: C1qtnf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204152.1 | NP_001191081.1 | 2, 3 | 373 | ||
NM_028331.5 | NP_082607.3 | 2, 3 | 373 | |||
GenBank mRNA | AK012868.1 | - | 2, 3 | 315 | ||
AK051058.1 | - | 2, 3 | 287 | |||
AK154407.1 | - | 2, 3 | 350 | |||
BC071187.1 | - | 2, 3 | 340 | 6395221 | ||
DQ002399.1 | - | 2, 3 | 339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
