siRNA Details
Gene Symbol: MYBPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000256.3 | NP_000247.2 | 18 | 1721 | ||
GenBank mRNA | AK291569.1 | - | 18 | 1721 | ||
AK297843.1 | - | 18 | 1721 | |||
BC136543.1 | - | 18 | 1698 | 9020533 | ||
BC136546.1 | - | 18 | 1698 | 9020536 | ||
BC142685.1 | - | 18 | 1701 | 40148564 | ||
BC151211.1 | - | 18 | 1704 | 40147517 | ||
EF560722.1 | - | 18 | 1722 | |||
EU747718.1 | - | 18 | 1721 | |||
EU747719.1 | - | 18 | 1721 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
