siRNA Details
Gene Symbol: Eldr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013290.1 | - | 3 | 206 | ||
AK015657.1 | - | 4 | 477 | |||
AK165613.1 | - | 4 | 307 | |||
BC037766.1 | - | 3 | 181 | |||
BC054406.1 | - | 3 | 125 | |||
BI990615.1 | - | 5 | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
