siRNA Details
Gene Symbol: MYBPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK291569.1 | - | 30 | 3333 | ||
BC142685.1 | - | 30 | 3313 | 40148564 | ||
BC151211.1 | - | 30 | 3316 | 40147517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
