siRNA Details
Gene Symbol: Hdhd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024257.1 | NP_077219.1 | 2 | 612 | ||
GenBank mRNA | AK013245.1 | - | 2 | 627 | ||
BC003491.1 | - | 2 | 612 | 2811559 |

More Information
Other Species Targeted

Gene Ontology Categories:
