siRNA Details
Gene Symbol: Tonsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183091.3 | NP_898914.3 | 23 | 3716 | ||
GenBank mRNA | AK013272.1 | - | 3 | 514 | ||
BC066068.1 | - | 23 | 3698 | 6851399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
