siRNA Details
Gene Symbol: Sass6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289568.1 | NP_001276497.1 | 13 | 1578 | ||
NM_001289571.1 | NP_001276500.1 | 11 | 1434 | |||
NM_028349.3 | NP_082625.2 | 12 | 1495 | |||
GenBank mRNA | AK013338.1 | - | 13 | 1470 | ||
AK165224.1 | - | 13 | 1524 | |||
BC050110.1 | - | 12 | 1445 | 30024366 |
