siRNA Details
Gene Symbol: Veph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145820.3 | NP_665819.2 | 12 | 2496 | ||
GenBank mRNA | AB085187.1 | - | 12 | 2478 | ||
AB085189.1 | - | 4 | 541 | |||
AK013398.1 | - | 4 | 556 | |||
BC075649.1 | - | 12 | 2496 | |||
BC094339.1 | - | 4 | 509 | |||
BC128271.1 | - | 11 | 2203 | 40059997 |
