siRNA Details
Gene Symbol: Fam76b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176836.3 | NP_789806.1 | 7, 8 | 941 | ||
GenBank mRNA | AK052430.1 | - | 7, 8 | 900 | ||
AK145026.1 | - | 7, 8 | 956 | |||
BC043120.1 | - | 7, 8 | 941 | 6408908 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006242531, XM_006242529, XM_006242526.
Gene Ontology Categories:
