siRNA Details
Gene Symbol: Kctd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289671.1 | NP_001276600.1 | 3 | 447 | ||
NM_001289672.1 | NP_001276601.1 | 3 | 447 | |||
NM_001289673.1 | NP_001276602.1 | 3 | 447 | |||
GenBank mRNA | AK179755.1 | - | 2 | 135 | ||
BC144818.1 | - | 3 | 447 | |||
BC144820.1 | - | 3 | 447 | |||
BC145799.1 | - | 3 | 447 | 40131102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
