siRNA Details
Gene Symbol: Cxx1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028375.3 | NP_082651.1 | 1 | 272 | ||
GenBank mRNA | AK013603.1 | - | 1 | 163 | ||
BC031701.1 | - | 1 | 160 | 3594379 | ||
BY123946.1 | - | 1 | 272 | |||
CK627323.1 | - | 1 | 189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
