siRNA Details
Gene Symbol: Setd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028385.1 | NP_082661.1 | 10 | 1322 | ||
GenBank mRNA | AK013649.1 | - | 6 | 1252 | ||
AK049143.1 | - | 4 | 400 | |||
AK122551.1 | - | 10 | 1322 | |||
AK164036.1 | - | 5 | 3227 | |||
BC056502.1 | - | 12 | 1858 | |||
BC083184.1 | - | 9 | 1203 | 30536831 |
