siRNA Details
Gene Symbol: Macrod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028387.1 | NP_082663.1 | 3, 4 | 508 | ||
GenBank mRNA | AK004072.1 | - | 3, 4 | 465 | ||
AK011824.1 | - | 3, 4 | 506 | |||
AK029184.1 | - | 3, 4 | 488 | |||
BC038317.1 | - | 3, 4 | 434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
