siRNA Details
Gene Symbol: Hepacam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175189.4 | NP_780398.2 | 5 | 940 | ||
GenBank mRNA | AK013637.1 | - | 5 | 940 | ||
AK081154.1 | - | 5 | 856 | |||
AK138438.1 | - | 4 | 464 | |||
AK158265.1 | - | 5 | 1178 | |||
BC082537.1 | - | 5 | 854 | |||
BC139057.1 | - | 5 | 930 | 8862079 | ||
BC139058.1 | - | 5 | 930 | 8862080 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
