siRNA Details
Gene Symbol: Glrx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028419.2 | NP_082695.1 | 1, 2 | 292 | ||
GenBank mRNA | AK009158.1 | - | 1, 2 | 297 | ||
AK013761.1 | - | 1, 2 | 293 | |||
AK050883.1 | - | 292 | ||||
BC050937.1 | - | 1, 2 | 312 | |||
BC058371.1 | - | 1, 2 | 312 | 6819353 | ||
DQ083330.1 | - | 1, 2 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
