siRNA Details
Gene Symbol: Fut11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028428.2 | NP_082704.1 | 2 | 1053 | ||
GenBank mRNA | AJ542393.1 | - | 2 | 1052 | ||
AK014029.1 | - | 2 | 1037 | |||
AK029717.1 | - | 2 | 1053 | |||
AK030310.1 | - | 2 | 1052 | |||
AK032390.1 | - | 1 | 1135 | |||
AK046461.1 | - | 2 | 1053 | |||
BC138555.1 | - | 2 | 1052 | 8861575 | ||
BC145735.1 | - | 2 | 1052 | 40131038 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
