siRNA Details
Gene Symbol: Ppil6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028430.1 | NP_082706.1 | 1, 2 | 178 | ||
GenBank mRNA | AK013818.1 | - | 1, 2 | 141 | ||
BC147756.1 | - | 1, 2 | 174 | 9056705 | ||
BC147757.1 | - | 1, 2 | 174 | 9056706 | ||
BY104076.1 | - | 1, 2 | 179 | |||
CA466046.1 | - | 1, 2 | 160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
