siRNA Details
Gene Symbol: 2900092C05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028434.3 | NP_082710.2 | 5 | 424 | ||
GenBank mRNA | AK013837.1 | - | 5 | 440 | ||
BC087883.1 | - | 5 | 424 | 6772737 | ||
BC099547.1 | - | 5 | 427 | 6741066 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
