siRNA Details
Gene Symbol: 3110009E18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172074.1 | NP_001165545.1 | 3, 4 | 813 | ||
NM_028439.2 | NP_082715.1 | 3, 4 | 220 | |||
GenBank mRNA | AK014026.1 | - | 3, 4 | 814 | ||
BC028767.1 | - | 3, 4 | 212 | 1264217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
