siRNA Details
Gene Symbol: Fam101a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028443.2 | NP_082719.1 | 2, 3 | 374 | ||
GenBank mRNA | AK014112.1 | - | 2, 3 | 404 | ||
BC053444.1 | - | 2, 3 | 374 | 6336076 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
