siRNA Details
Gene Symbol: Cenpv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028448.1 | NP_082724.1 | 4 | 588 | ||
GenBank mRNA | AK014046.1 | - | 4 | 588 | ||
BC038868.1 | - | 1 | 303 | |||
BC147794.1 | - | 4 | 550 | 9087511 | ||
BC147795.1 | - | 4 | 550 | 9087512 | ||
BC147835.1 | - | 4 | 550 | 9087552 | ||
BC147837.1 | - | 4 | 550 | 9087554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
