siRNA Details
Gene Symbol: Pcdh18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130448.3 | NP_569715.3 | 1 | 2004 | ||
GenBank mRNA | AF416735.1 | - | 1 | 1949 | ||
AK014140.1 | - | 1 | 152 | |||
AK032820.1 | - | 1 | 1982 | |||
AK036560.1 | - | 1 | 1973 | |||
AK173204.1 | - | 1 | 1949 | |||
BC052198.1 | - | 1 | 1968 | 6512513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
