siRNA Details
Gene Symbol: Pcdh18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF416735.1 | - | 3 | 3035 | ||
AK173204.1 | - | 3 | 3035 | |||
BC052198.1 | - | 3 | 3054 | 6512513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
