siRNA Details
Gene Symbol: Tbkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198100.2 | NP_932768.2 | 1, 2 | 358 | ||
GenBank mRNA | AK014173.1 | - | 2, 3 | 320 | ||
BC057590.1 | - | 1, 2 | 358 | 6810986 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
