siRNA Details
Gene Symbol: 3110052M02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166497.1 | NP_001159969.1 | 6 | 1317 | ||
GenBank mRNA | AB010359.1 | - | 1 | 50 | ||
AK161479.1 | - | 6 | 2663 | |||
BC064464.1 | - | 3 | 1137 |
Gene Symbol: Gm10509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB010360.1 | - | 1 | 50 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
