siRNA Details
Gene Symbol: 3110052M02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166497.1 | NP_001159969.1 | 6 | 1551 | ||
GenBank mRNA | AK155999.1 | - | 6 | 1539 | ||
AK161479.1 | - | 6 | 2897 | |||
BC064464.1 | - | 3 | 1371 |
