siRNA Details
Gene Symbol: Mrgbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028479.1 | NP_082755.1 | 3 | 672 | ||
GenBank mRNA | AK005551.1 | - | 2 | 312 | ||
AK052575.1 | - | 2 | 327 | |||
AK217601.1 | - | 1 | 89 | |||
BC042508.1 | - | 2 | 286 | |||
BC054431.1 | - | 2 | 259 | |||
BC085320.1 | - | 1 | 778 | |||
BC110993.1 | - | 2 | 281 | 5363496 | ||
BC116732.1 | - | 3 | 672 | 40126051 | ||
BC116734.1 | - | 3 | 672 | 40126053 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
