siRNA Details
Gene Symbol: Cib4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028483.1 | NP_082759.1 | 4 | 293 | ||
GenBank mRNA | AK006670.1 | - | 4 | 294 | ||
BC099558.1 | - | 4 | 239 | |||
BC118938.1 | - | 4 | 258 | 40091772 |
