siRNA Details
Gene Symbol: Gpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122963.1 | NP_001116435.1 | 7 | 1845 | ||
NM_028487.4 | NP_082763.3 | 8 | 1905 | |||
GenBank mRNA | AK052018.1 | - | 1 | 1331 | ||
AK082704.1 | - | 7 | 1663 | |||
AK135728.1 | - | 8 | 1836 | |||
AK136894.1 | - | 8 | 1684 | |||
AK145318.1 | - | 7 | 1834 | |||
AK151283.1 | - | 6 | 1066 | |||
AK161928.1 | - | 7 | 1633 | |||
AK163370.1 | - | 8 | 1753 | |||
AK163390.1 | - | 8 | 1907 | |||
AK169406.1 | - | 8 | 1700 | |||
AY382529.1 | - | 8 | 1703 | |||
BC016083.1 | - | 6 | 699 | 4501399 | ||
BC067075.1 | - | 7 | 1619 | 30614988 | ||
BC076635.1 | - | 7 | 1675 | |||
BC094888.1 | - | 7 | 1638 | 30628044 | ||
BC096442.1 | - | 8 | 1720 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
