siRNA Details
Gene Symbol: Ccdc132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167750.1 | NP_001161222.1 | 16, 17 | 1488 | ||
NM_001167751.1 | NP_001161223.1 | 16, 17 | 1488 | |||
NM_024260.5 | NP_077222.4 | 16, 17 | 1488 | |||
GenBank mRNA | AK042031.1 | - | 16, 17 | 1468 | ||
AK075898.1 | - | 16, 17 | 1477 | |||
AK129458.1 | - | 16, 17 | 1438 | |||
AY254697.1 | - | 16, 17 | 1481 | |||
BC002304.1 | - | 1, 2 | 33 | 3591600 | ||
BC036294.1 | - | 16, 17 | 1428 | 3500316 | ||
BC043092.1 | - | 16, 17 | 1445 | 6405356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
