siRNA Details
Gene Symbol: Calr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028500.3 | NP_082776.2 | 5 | 687 | ||
NM_029782.3 | NP_084058.2 | 3 | 388 | |||
GenBank mRNA | AB206786.1 | - | 5 | 592 | ||
AK006582.1 | - | 5 | 688 | |||
AK018263.1 | - | 3 | 310 | |||
AK046081.1 | - | 3 | 389 |

More Information
Other Species Targeted

Gene Ontology Categories:
