siRNA Details
Gene Symbol: Clhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163521.1 | NP_001156993.1 | 5 | 857 | ||
NM_028504.1 | NP_082780.1 | 5 | 890 | |||
GenBank mRNA | AK006591.1 | - | 5 | 891 | ||
BC137683.1 | - | 5 | 814 | 8860702 | ||
BC144828.1 | - | 5 | 781 | 9053376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
