siRNA Details
Gene Symbol: 1700034J05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164236.1 | NP_001157708.1 | 2 | 475 | ||
NM_028509.1 | NP_082785.1 | 2 | 475 | |||
GenBank mRNA | AK006598.1 | - | 2 | 468 | ||
AK029686.1 | - | 462 | ||||
AK076643.1 | - | 2 | 469 | |||
BC052362.1 | - | 2 | 467 | 6707822 |
