siRNA Details
Gene Symbol: Col20a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028518.1 | NP_082794.1 | 22, 23 | 2908 | ||
GenBank mRNA | BC030415.1 | - | 10, 11 | 1243 |

More Information
Other Species Targeted

Gene Ontology Categories:
