siRNA Details
Gene Symbol: Tex33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163612.2 | NP_001157084.1 | 6 | 1007 | ||
NM_001163613.1 | NP_001157085.1 | 5 | 802 | |||
NM_028522.1 | NP_082798.1 | 7 | 932 | |||
GenBank mRNA | AK006856.1 | - | 7 | 906 | ||
BC100472.1 | - | 7 | 858 | |||
BC107197.2 | - | 6 | 811 | 40051085 | ||
BC107198.1 | - | 6 | 811 | 40051089 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
