siRNA Details
Gene Symbol: Dcbld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028523.3 | NP_082799.2 | 9 | 1323 | ||
GenBank mRNA | AF387548.1 | - | 9 | 1415 | ||
AK051889.1 | - | 7 | 601 | |||
BC066097.1 | - | 9 | 1323 | 6856458 |
